Assay Details
| Gene Symbol: EGLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928507.6 | XP_001928542.3 | 1019 | 78 | ||
| GenBank mRNA | AK394727.1 | - | 5 - 6 | 544 | 78 | |
More Information
Set Membership: |
|

EGLN3
Genomic Map