Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096634.1 | XP_020952293.1 | 4006 | 99 | ||
| XM_021096635.1 | XP_020952294.1 | 4006 | 99 | |||
| XM_021096636.1 | XP_020952295.1 | 4006 | 99 | |||
| XM_021096637.1 | XP_020952296.1 | 4006 | 99 | |||
| XM_021096638.1 | XP_020952297.1 | 4006 | 99 | |||
| XM_021096639.1 | XP_020952298.1 | 4006 | 99 | |||
| XM_021096640.1 | XP_020952299.1 | 3793 | 99 | |||
| XM_021096641.1 | XP_020952300.1 | 3793 | 99 | |||
| XM_021096642.1 | XP_020952301.1 | 3793 | 99 | |||
| XM_021096643.1 | XP_020952302.1 | 3793 | 99 | |||
| XM_021096644.1 | XP_020952303.1 | 3673 | 99 | |||
| XM_021096645.1 | XP_020952304.1 | 3673 | 99 | |||
| GenBank mRNA | AK394735.1 | - | 4 - 5 | 410 | 99 | |
More Information
Set Membership: |
|

TUT4
Genomic Map