Assay Details
| Gene Symbol: RAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133878.4 | XP_003133926.1 | 400 | 73 | ||
| XM_013984735.2 | XP_013840189.1 | 426 | 73 | |||
| GenBank mRNA | AK394811.1 | - | 2 - 3 | 359 | 73 | |
More Information
Set Membership: |
|

RAD1
Genomic Map