Assay Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125363.6 | XP_003125411.1 | 1465 | 70 | ||
| GenBank mRNA | AK394826.1 | - | 3 - 4 | 526 | 70 | |
More Information
Set Membership: |
|

TP53I3
Genomic Map