Assay Details
| Gene Symbol: LOC100516852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086088.1 | XP_020941747.1 | 2766 | 100 | ||
| GenBank mRNA | AK394969.1 | - | 8 - 9 | 1054 | 100 | |
More Information
Set Membership: |
|

LOC100516852
Genomic Map