Assay Details
| Gene Symbol: DELE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124042.6 | XP_003124090.3 | 3775 | 116 | ||
| GenBank mRNA | AK395002.1 | - | 6 - 7 | 801 | 116 | |
More Information
Set Membership: |
|

DELE1
Genomic Map