Assay Details
| Gene Symbol: KIF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077423.1 | XP_020933082.1 | 5539 | 110 | ||
| XM_021077424.1 | XP_020933083.1 | 5487 | 110 | |||
| XM_021077425.1 | XP_020933084.1 | 5752 | 110 | |||
| GenBank mRNA | AK395117.1 | - | 1 | 2020 | 110 | |
More Information
Set Membership: |
|

KIF3B
Genomic Map