Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094952.1 | XP_020950611.1 | 2072 | 104 | ||
| XM_021094959.1 | XP_020950618.1 | 2070 | 104 | |||
| XM_021094972.1 | XP_020950631.1 | 2072 | 104 | |||
| GenBank mRNA | AK395248.1 | - | 1 - 2 | 98 | 104 | |
More Information
Set Membership: |
|

DNAAF4
Genomic Map