Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013997511.2 | XP_013852965.2 | 7217 | 104 | ||
| XM_021091473.1 | XP_020947132.1 | 3713 | 104 | |||
| GenBank mRNA | AK395262.1 | - | 1 | 1917 | 104 | |
More Information
Set Membership: |
|

MGAT3
Genomic Map