Assay Details
| Gene Symbol: KIAA0319L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095911.1 | XP_020951570.1 | 407 | 78 | ||
| XM_021095912.1 | XP_020951571.1 | 310 | 78 | |||
| XM_021095913.1 | XP_020951572.1 | 246 | 78 | |||
| GenBank mRNA | AK395524.1 | - | 2 - 3 | 362 | 78 | |
More Information
Set Membership: |
|

KIAA0319L
Genomic Map