Assay Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657908.3 | XP_005657965.2 | 1325 | 111 | ||
| GenBank mRNA | AK395550.1 | - | 2 - 3 | 229 | 111 | |
More Information
Set Membership: |
|

MID2
Genomic Map