Assay Details
| Gene Symbol: YAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062706.1 | XP_020918365.1 | 692 | 69 | ||
| GenBank mRNA | AK395568.1 | - | 3 - 4 | 1105 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: YAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062706.1 | XP_020918365.1 | 692 | 69 | ||
| GenBank mRNA | AK395568.1 | - | 3 - 4 | 1105 | 69 | |
Set Membership: |
|