Assay Details
Gene Symbol: SPRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927097.5 | XP_001927132.1 | 503 | 86 | ||
GenBank mRNA | AK395766.1 | - | 1 - 2 | 514 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001927097.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
