Assay Details
| Gene Symbol: INO80E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088006.1 | XP_020943665.1 | 284 | 66 | ||
| XM_021088007.1 | XP_020943666.1 | 284 | 66 | |||
| XM_021088008.1 | XP_020943667.1 | 284 | 66 | |||
| XM_021088009.1 | XP_020943668.1 | 284 | 66 | |||
| GenBank mRNA | AK396186.1 | - | 2 - 3 | 225 | 66 | |
More Information
Set Membership: |
|

INO80E
Genomic Map