Assay Details
| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081503.1 | XP_020937162.1 | 109 | |||
| GenBank mRNA | AK396319.1 | - | 206 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081503.1 | XP_020937162.1 | 109 | |||
| GenBank mRNA | AK396319.1 | - | 206 | 109 | ||
Set Membership: |
|