Assay Details
| Gene Symbol: ELF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130998.6 | XP_003131046.3 | 1499 | 67 | ||
| XM_005668448.3 | XP_005668505.1 | 1394 | 67 | |||
| XM_021065634.1 | XP_020921293.1 | 1219 | 67 | |||
| XM_021065635.1 | XP_020921294.1 | 1294 | 67 | |||
| XM_021065636.1 | XP_020921295.1 | 2491 | 67 | |||
| XM_021065637.1 | XP_020921296.1 | 1122 | 67 | |||
| XM_021065638.1 | XP_020921297.1 | 1172 | 67 | |||
| XM_021065639.1 | XP_020921298.1 | 2385 | 67 | |||
| GenBank mRNA | AK396320.1 | - | 5 - 6 | 859 | 67 | |
More Information
Set Membership: |
|

ELF1
Genomic Map