Assay Details
| Gene Symbol: EBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130959.5 | XP_003131007.1 | 255 | 81 | ||
| XM_021065312.1 | XP_020920971.1 | 279 | 81 | |||
| GenBank mRNA | AK396354.1 | - | 1 - 2 | 209 | 81 | |
More Information
Set Membership: |
|

EBPL
Genomic Map