Assay Details
| Gene Symbol: CHST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135049.4 | XP_003135097.2 | 1715 | 108 | ||
| GenBank mRNA | AK396402.1 | - | 1 - 2 | 1686 | 108 | |
More Information
Set Membership: |
|

CHST7
Genomic Map