Assay Details
| Gene Symbol: SYNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356291.5 | XP_003356339.1 | 148 | 93 | ||
| XM_005665184.3 | XP_005665241.1 | 148 | 93 | |||
| XM_005665185.3 | XP_005665242.1 | 148 | 93 | |||
| XM_013999124.2 | XP_013854578.1 | 148 | 93 | |||
| GenBank mRNA | AK396421.1 | - | 1 - 2 | 89 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003356291.5, XM_005665184.3, XM_005665185.3, XM_013999124.2 transcripts.
Set Membership: |
|

SYNC
Genomic Map