Assay Details
| Gene Symbol: LOC100154105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924950.6 | XP_001924985.5 | 3108 | 107 | ||
| GenBank mRNA | AK396439.1 | - | 1 | 1724 | 107 | |
More Information
Set Membership: |
|

LOC100154105
Genomic Map