Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482141.4 | XP_003482189.1 | 1612 | 105 | ||
| XM_021099793.1 | XP_020955452.1 | 1626 | 105 | |||
| GenBank mRNA | AK396441.1 | - | 6 - 7 | 1302 | 105 | |
More Information
Set Membership: |
|

PGBD1
Genomic Map