Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984310.2 | XP_013839764.1 | 702 | 104 | ||
| XM_013984313.2 | XP_013839767.1 | 1589 | 104 | |||
| XM_021075639.1 | XP_020931298.1 | 5611 | 104 | |||
| XM_021075640.1 | XP_020931299.1 | 1533 | 104 | |||
| GenBank mRNA | AK396542.1 | - | 5 - 6 | 1126 | 104 | |
More Information
Set Membership: |
|

ZNF385B
Genomic Map