Assay Details
| Gene Symbol: ZNF404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655939.3 | XP_005655996.1 | 2139 | 84 | ||
| GenBank mRNA | AK396752.1 | - | 1 | 1813 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ZNF404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655939.3 | XP_005655996.1 | 2139 | 84 | ||
| GenBank mRNA | AK396752.1 | - | 1 | 1813 | 84 | |
Set Membership: |
|