Assay Details
| Gene Symbol: CMTM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071540.1 | XP_020927199.1 | 2725 | 76 | ||
| GenBank mRNA | AK396885.1 | - | 1 | 1419 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CMTM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071540.1 | XP_020927199.1 | 2725 | 76 | ||
| GenBank mRNA | AK396885.1 | - | 1 | 1419 | 76 | |
Set Membership: |
|