Assay Details
| Gene Symbol: GNAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021072293.1 | XP_020927952.1 | 1340 | 63 | ||
| GenBank mRNA | AK396931.1 | - | 1 - 2 | 549 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GNAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021072293.1 | XP_020927952.1 | 1340 | 63 | ||
| GenBank mRNA | AK396931.1 | - | 1 - 2 | 549 | 63 | |
Set Membership: |
|