Assay Details
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129269.4 | XP_003129317.1 | 977 | 101 | ||
| GenBank mRNA | AK396941.1 | - | 1 - 2 | 471 | 101 | |
More Information
Set Membership: |
|

DKK2
Genomic Map