Assay Details
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129269.4 | XP_003129317.1 | 1129 | 86 | ||
| GenBank mRNA | AK396941.1 | - | 2 - 3 | 623 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129269.4 | XP_003129317.1 | 1129 | 86 | ||
| GenBank mRNA | AK396941.1 | - | 2 - 3 | 623 | 86 | |
Set Membership: |
|