Assay Details
| Gene Symbol: SLC40A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483701.4 | XP_003483749.1 | 1747 | 69 | ||
| XM_013984335.2 | XP_013839789.1 | 1488 | 69 | |||
| GenBank mRNA | AK397115.1 | - | 2 - 3 | 698 | 69 | |
More Information
Set Membership: |
|

SLC40A1
Genomic Map