Assay Details
| Gene Symbol: SDR9C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355478.4 | XP_003355526.4 | 8093 | 75 | ||
| XM_021091808.1 | XP_020947467.1 | 6959 | 75 | |||
| GenBank mRNA | AK397372.1 | - | 2 - 3 | 476 | 75 | |
More Information
Set Membership: |
|

SDR9C7
Genomic Map