Assay Details
| Gene Symbol: CDK5RAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067091.1 | XP_020922750.1 | 1573 | 66 | ||
| GenBank mRNA | AK397598.1 | - | 7 - 8 | 945 | 66 | |
More Information
Set Membership: |
|

CDK5RAP3
Genomic Map