Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661403.2 | XP_005661460.1 | 2077 | 85 | ||
| XM_013994828.2 | XP_013850282.1 | 2047 | 85 | |||
| XM_013994830.2 | XP_013850284.1 | 1858 | 85 | |||
| XM_013994831.1 | XP_013850285.1 | 1836 | 85 | |||
| XM_013994832.1 | XP_013850286.1 | 1781 | 85 | |||
| XM_021084240.1 | XP_020939899.1 | 1979 | 85 | |||
| XM_021084241.1 | XP_020939900.1 | 1991 | 85 | |||
| XM_021084242.1 | XP_020939901.1 | 1975 | 85 | |||
| XM_021084244.1 | XP_020939903.1 | 2485 | 85 | |||
| XM_021084245.1 | XP_020939904.1 | 1889 | 85 | |||
| XM_021084246.1 | XP_020939905.1 | 2031 | 85 | |||
| XM_021084247.1 | XP_020939906.1 | 3350 | 85 | |||
| GenBank mRNA | AK397689.1 | - | 1 - 2 | 318 | 85 | |
More Information
Set Membership: |
|

PWWP3A
Genomic Map