Assay Details
| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076637.1 | XP_020932296.1 | 4559 | 117 | ||
| XM_021076638.1 | XP_020932297.1 | 2269 | 117 | |||
| GenBank mRNA | AK397700.1 | - | 2 - 3 | 2478 | 117 | |
More Information
Set Membership: |
|

INSYN2B
Genomic Map