Assay Details
| Gene Symbol: LOC100524016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124695.4 | XP_003124743.2 | 1343 | 76 | ||
| GenBank mRNA | AK397860.1 | - | 9 - 10 | 1279 | 76 | |
More Information
Set Membership: |
|

LOC100524016
Genomic Map