Assay Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089073.1 | XP_020944732.1 | 1670 | 64 | ||
| XM_021089074.1 | XP_020944733.1 | 55 | ||||
| XM_021089075.1 | XP_020944734.1 | 1342 | 64 | |||
| GenBank mRNA | AK398327.1 | - | 7 - 8 | 696 | 64 | |
More Information
Set Membership: |
|

PTDSS1
Genomic Map