Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129788.5 | XP_003129836.2 | 2019 | 71 | ||
| GenBank mRNA | AK398485.1 | - | 5 - 6 | 605 | 71 | |
More Information
Set Membership: |
|

MRE11
Genomic Map