Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065041.1 | XP_020920700.1 | 9223 | 70 | ||
| XM_021065042.1 | XP_020920701.1 | 9202 | 70 | |||
| XM_021065043.1 | XP_020920702.1 | 9058 | 70 | |||
| XM_021065044.1 | XP_020920703.1 | 7737 | 70 | |||
| XM_021065045.1 | XP_020920704.1 | 7980 | 70 | |||
| XM_021065046.1 | XP_020920705.1 | 7781 | 70 | |||
| XM_021065047.1 | XP_020920706.1 | 8084 | 70 | |||
| XM_021065048.1 | XP_020920707.1 | 7992 | 70 | |||
| XM_021065049.1 | XP_020920708.1 | 8564 | 70 | |||
| XM_021065050.1 | XP_020920709.1 | 8317 | 70 | |||
| XM_021065051.1 | XP_020920710.1 | 7740 | 70 | |||
| XM_021065052.1 | XP_020920711.1 | 7277 | 70 | |||
| GenBank mRNA | AK398495.1 | - | 2 - 3 | 250 | 70 | |
More Information
Set Membership: |
|

TASOR2
Genomic Map