Assay Details
| Gene Symbol: DNER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076495.1 | XP_020932154.1 | 1405 | 54 | ||
| GenBank mRNA | AK398503.1 | - | 3 - 4 | 767 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DNER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076495.1 | XP_020932154.1 | 1405 | 54 | ||
| GenBank mRNA | AK398503.1 | - | 3 - 4 | 767 | 54 | |
Set Membership: |
|