Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134669.5 | XP_003134717.3 | 1181 | 69 | ||
| GenBank mRNA | AK398573.1 | - | 2 - 3 | 237 | 69 | |
More Information
Set Membership: |
|

PODXL
Genomic Map