Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065899.1 | XP_020921558.1 | 1056 | 82 | ||
| GenBank mRNA | AK398977.1 | - | 3 - 4 | 361 | 82 | |
More Information
Set Membership: |
|

POGLUT2
Genomic Map