Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065899.1 | XP_020921558.1 | 1684 | 168 | ||
| GenBank mRNA | AK398977.1 | - | 6 - 7 | 989 | 168 | |
More Information
Set Membership: |
|

POGLUT2
Genomic Map