Assay Details
| Gene Symbol: ARHGAP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070329.1 | XP_020925988.1 | 960 | 80 | ||
| XM_021070330.1 | XP_020925989.1 | 961 | 80 | |||
| GenBank mRNA | AK399008.1 | - | 2 - 3 | 879 | 80 | |
| AK400704.1 | - | 2 - 3 | 965 | 80 | ||
More Information
Set Membership: |
|

ARHGAP31
Genomic Map