Assay Details
| Gene Symbol: RFLNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926661.5 | XP_001926696.2 | 1783 | 66 | ||
| XM_005670568.2 | XP_005670625.1 | 1554 | 66 | |||
| XM_013982774.2 | XP_013838228.1 | 6110 | 66 | |||
| XM_021072253.1 | XP_020927912.1 | 1906 | 66 | |||
| GenBank mRNA | AK399069.1 | - | 1 | 821 | 66 | |
More Information
Set Membership: |
|

RFLNA
Genomic Map