Assay Details
| Gene Symbol: SRSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927419.6 | XP_001927454.2 | 597 | 84 | ||
| XM_001927434.6 | XP_001927469.2 | 452 | 84 | |||
| XM_005666331.3 | XP_005666388.1 | 455 | 84 | |||
| XM_005666334.3 | XP_005666391.1 | 558 | 84 | |||
| XM_005666335.2 | XP_005666392.1 | 466 | 84 | |||
| XM_005666336.3 | XP_005666393.1 | 452 | 84 | |||
| XM_021099406.1 | XP_020955065.1 | 895 | 84 | |||
| XM_021099407.1 | XP_020955066.1 | 895 | 84 | |||
| GenBank mRNA | AK399777.1 | - | 4 - 5 | 425 | 84 | |
More Information
Set Membership: |
|

SRSF5
Genomic Map