Assay Details
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122986.4 | XP_003123034.2 | 717 | 58 | ||
| GenBank mRNA | AK399778.1 | - | 4 - 5 | 627 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122986.4 | XP_003123034.2 | 717 | 58 | ||
| GenBank mRNA | AK399778.1 | - | 4 - 5 | 627 | 58 | |
Set Membership: |
|