Assay Details
| Gene Symbol: GLT8D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925457.5 | XP_001925492.3 | 1365 | 68 | ||
| XM_005669641.3 | XP_005669698.2 | 3802 | 68 | |||
| XM_021068949.1 | XP_020924608.1 | 5896 | 68 | |||
| GenBank mRNA | AY609623.1 | - | 7 - 8 | 846 | 68 | |
More Information
Set Membership: |
|

GLT8D1
Genomic Map