Assay Details
| Gene Symbol: TCF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087668.1 | XP_020943327.1 | 740 | 87 | ||
| GenBank mRNA | AY609724.1 | - | 1 - 2 | 456 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TCF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087668.1 | XP_020943327.1 | 740 | 87 | ||
| GenBank mRNA | AY609724.1 | - | 1 - 2 | 456 | 87 | |
Set Membership: |
|