Assay Details
| Gene Symbol: CCT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662018.3 | XP_005662075.1 | 837 | 76 | ||
| GenBank mRNA | AY610017.1 | - | 6 - 7 | 791 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662018.3 | XP_005662075.1 | 837 | 76 | ||
| GenBank mRNA | AY610017.1 | - | 6 - 7 | 791 | 76 | |
Set Membership: |
|