Assay Details
| Gene Symbol: MED30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135161.4 | XP_003135209.1 | 496 | 94 | ||
| XM_013996532.2 | XP_013851986.1 | 498 | 94 | |||
| GenBank mRNA | AY610104.1 | - | 2 - 3 | 474 | 94 | |
More Information
Set Membership: |
|

MED30
Genomic Map