Assay Details
| Gene Symbol: LRRC39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090152.1 | XP_020945811.1 | 1151 | 83 | ||
| XM_021090153.1 | XP_020945812.1 | 902 | 83 | |||
| GenBank mRNA | AY610221.1 | - | 6 - 7 | 870 | 83 | |
More Information
Set Membership: |
|

LRRC39
Genomic Map