Assay Details
| Gene Symbol: RPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021102423.1 | XP_020958082.1 | 294 | 113 | ||
| GenBank mRNA | AY610313.1 | - | 1 - 2 | 142 | 113 | |
More Information
Set Membership: |
|

RPA3
Genomic Map